Series 2000 Benchtop Meters, Environmental Express
Supplier: Antyila Scientific
35419-54 35419-55 35419-60 35419-62 35419-65 35419-67 35419-70 35419-72
76595-366EA 1494.16 CAD
76595-366 76595-368 76595-370 76595-372 76595-374 76595-376 76595-378 76595-380
Series 2000 Benchtop Meters, Environmental Express
Electrochemistry Meters pH and Multi-Parameter Meters
Developed using extensive feedback from laboratory staff, our new Environmental Express 2000 series meters are assembled in various configurations for measuring a pH/mV, EC, Ion, DO, BOD parameters.
  • Various configurations available to meet the demands of measuring pH/mV, EC, Ion, PC, PD, DO, BOD parameters
  • Automatic record retention and accessibility for continuous compliance with regulatory requirements
  • 0.01, 0.001 resolution for routine or research applications where acute accuracy is critical
  • Large displays with a small footprint, ideal for labs with limited bench space
  • Automatic functions like buffer recognition and auto calibration for ease of use at any skill level
Environmental Express/Oakton provides a diverse range of water quality analyzers and electrodes that are designed specifically for laboratory applications with compliance in mind. Our multi-parameter benchtops include functionality to meet the highly specific and/or broad ranging needs of wastewater, drinking water, food and beverage, chemical and pharmaceutical testing labs.
These benchtop meters offer a high-capacity internal memory that can capture and store 2000 data points with the time and date. They have a time-based auto data log with GLP compliant real time clocks. This data can be exported into CSV, Excel, or PDF formats via USB cable and use of complimentary software. A password access protection function secures the meter setup mode from unauthorized access. An adjustable shutoff time up to 30 minutes turns the meter off when idle.
Ordering information: Available with or without an electrode stand. Three-year warranty.