Sol-Vet® IV Catheters (Veterinary Care), Sol M
Supplier: SOL-MILLENNIUM CANADA, INC. CA
110301050001 110301050002 110301050003 110301050004 110301050005 110301050006 110301050007
76582-486BX 112.62 CAD
76582-486 76582-488 76582-490 76582-492 76582-494 76582-496 76582-498
Sol-Vet® IV Catheters (Veterinary Care), Sol M
Catheters
The Sol-Vet I.V. Catheter is used for continuous and intermittent intravenous medication delivery. It is designed with a safety feature that covers the needle bevel, helping to prevent needlestick injuries.
  • Protective casing ensures safety of catheter during transportation
  • Translucent catheter hub and chamber allows for easy visualization of blood flashback
  • Contains a hydrophobic membrane filter to prevent blood leakage during insertion
  • Filter cap can be removed to allow connection with a syringe to assist insertion
Products for Veterinary use only.