Sol-Vet® Standard Insulin Syringes (Veterinary Care), Sol M
Supplier: SOL-MILLENNIUM CANADA, INC. CA
V5295 V5295-40 V5305 V5305-40 V12905 V12905-40 V1305 V1305-40 V32905-40 V3295 V3305 V3305-40
76582-954BX 31.92 CAD
76582-954 76582-956 76582-958 76582-960 76582-938 76582-940 76582-942 76582-944 76582-946 76582-948 76582-950 76582-952
Sol-Vet® Standard Insulin Syringes (Veterinary Care), Sol M
Syringes Syringes, Medical
The Sol-Vet Insulin Syringe provides accurate and comfortable insulin administration.
  • Bold and clear graduated insulin unit markings for accurate dosing
  • Low dead space - improved accuracy and less medication waste when compared to conventional syringes
  • Half unit markings
Products for veterinary use only.