Sol-Care® Safety Insulin Syringes, Sol M
Supplier: SOL-MILLENNIUM CANADA, INC. CA
100002IM 100017IM 100081IM 100089IM 100071IM
76582-508BX 66.14 CAD
76582-508 76582-514 76582-548 76582-556 76582-532
Sol-Care® Safety Insulin Syringes, Sol M
Syringes Syringes, Medical
This Insulin syringe provides accurate and comfortable insulin administration, with features to help prevent needlestick injuries.
  • Needles are tri-bevel and equipped with plunger lock and break feature that prevents reuse of syringe
  • Low dead space - improved accuracy and less medication waste when compared to conventional syringes
  • Bold and clear graduated insulin unit markings for accurate dosing
Safety feature: Needle retracts into the syringe barrel as a safety feature to prevent needlestick injuries.