Sol-M® Standard Syringes with Needle
Supplier: SOL-MILLENNIUM CANADA, INC. CA
1812010 1812015 1812110 1812210 181023 181025 1811815B 1852215 182025 1831815B 1832010 1832015 1832110 1832115 1832210 1832215 1832310 1832510 1832515 1832558 1851815B
76582-404BX 53.63 CAD
76582-404 76582-406 76582-408 76582-410 76582-398 76582-400 76582-644 76582-432 76582-402 76582-646 76582-412 76582-414 76582-416 76582-418 76582-420 76582-422 76582-424 76582-426 76582-428 76582-430 76582-654
Sol-M® Standard Syringes with Needle
Syringes Syringes, Medical
Standard needle and syringe combination.
  • High-transparency barrel to easily visualize contents
  • Silicone lubrication ensures smooth plunger movement
  • Plunger backstop that prevents accidental withdrawal of plunger from the barrel
  • Large finger grips allow for easy, controlled injections
The Sol-M syringe with needle features low dead space technology that helps reduce medication waste and improve dosing accuracy.