Sol Care® Safety Syringes with Exchangeable Needle, Sol M
Supplier: SOL-MILLENNIUM CANADA, INC. CA
140076IM 140077IM 160072IM 160076IM 100085IM 140072IM 100072IM 100074IM 100075IM 100076IM 100077IM 100078IM 100079IM
76582-590BX 81.02 CAD
76582-590 76582-592 76582-594 76582-596 76582-552 76582-588 76582-534 76582-536 76582-538 76582-540 76582-542 76582-544 76582-546
Sol Care® Safety Syringes with Exchangeable Needle, Sol M
Syringes Syringes, Medical
The Sol-Care safety syringe aids in the prevention of needlestick injuries by allowing the user to retract the needle into the barrel after medication delivery, covering the needle and locking it in place.
  • Compatible with all standard needles
  • Clear, bold graduations for accurate dosing
  • No Gap design allows the plunger to be pushed to the zero mark without activating the safety mechanism
Safety mechanism does not obstruct the view of the needle tip. Visual and audible confirmation of safety mechanism activation. Patented locking ring design – the syringe cannot be reused.