Sol-Care® IV Catheters with Protector Straight, Sol M
Supplier: SOL-MILLENNIUM MEDICAL
SIV004 SIV005 SIV006 SIV007 SIV008
76582-890BX 108.09 CAD
76582-890 76582-892 76582-894 76582-896 76582-898
Sol-Care® IV Catheters with Protector Straight, Sol M
Catheters
I.V. Catheter is used for continuous and intermittent intravenous medication delivery.
  • Passive safety mechanism for the prevention of needlestick injuries
  • Needle bevel is designed for easy insertion that minimizes patient discomfort
  • Catheter is made of polyurethane (PUR), allowing for a softer catheter insertion that improves patient comfort
  • Variety of catheter gauges and lengths to accommodate for different clinical specialties, including Neonatology
It is designed with a safety feature that covers the needle bevel, helping to prevent needlestick injuries.