Sol-M® Blunt Fill Needles
Supplier: SOL-MILLENNIUM CANADA, INC. CA
110021 110022 110022F
76582-354BX 21.95 CAD
76582-354 76582-356 76582-564
Sol-M® Blunt Fill Needles
Syringe Needles
The Sol-M Blunt Fill Needle reduces the risk of needlestick injuries to healthcare workers when drawing up and preparing medication by removing the need for sharps.
  • Eliminates the unnecessary use of sharps
  • Color-coded hubs for easy identification
  • Needle hub is compatible with all Luer Lock and Luer Slip Tip syringes
  • Available with 5-micron filter to prevent the drawing of glass particles when using ampules
This can be used with the ‘Double Needle Technique’ when drawing medication and can also be used for IV push administration in needleless IV systems.
Order Now
SPECIFICATIONS
Environmentally Preferable