Sol-Care® Safety Syringes without Needle, Sol M
Supplier: SOL-MILLENNIUM CANADA, INC. CA
120008IM 120009IM 120006IM 120007IM
76582-570BX 85.37 CAD
76582-570 76582-572 76582-566 76582-568
Sol-Care® Safety Syringes without Needle, Sol M
Syringes Syringes, Medical
Sol-Care Luer Lock Safety Syringes have a manual retractable needle mechanism, and is compatible with all hypodermic needles.
  • Compatible with all standard needles
  • Clear, bold graduations for accurate dosing
  • No Gap design allows the plunger to be pushed to the zero mark without activating the safety mechanism
The Sol-Care Safety Syringe aids in the prevention of needlestick injuries by allowing the user to retract the needle into the barrel after medication delivery, covering the needle and locking it in place. Safety mechanism does not obstruct the view of the needle tip. Visual and audible confirmation of safety mechanism activation.