Sol-Care® Safety Syringes with Fixed Needle, Sol M
Supplier: SOL-MILLENNIUM CANADA, INC. CA
100018IM 100019IM 100034IM 100040IM 100067IM 100082IM 100088IM 100090IM 100070IM
76582-510BX 64.56 CAD
76582-510 76582-512 76582-516 76582-518 76582-522 76582-524 76582-526 76582-550 76582-554 76582-558 76582-530
Sol-Care® Safety Syringes with Fixed Needle, Sol M
Syringes Syringes, Medical
The Sol-Care Safety Syringes are equipped with a fixed needle, preventing reuse and lowering the risk of needlestick injuries.
  • Convenient injection solution for key therapeutic areas, such as allergy and immunization
  • Clear, bold graduations for accurate dosing
  • No Gap design allows the plunger to be pushed to the zero mark without activating the safety mechanism
  • Visual and audible confirmation of safety mechanism activation
Low Dead Space design (0.01 ml) reduces waste and allows for more efficient medication delivery. The safety mechanism is activated when the user pulls the needle into the barrelof the syringe after medication delivery. This action covers the needle and locks prevention of needlestick injuries with a manual retractable needle mechanism. Patented locking ring design – the syringe cannot be reused once the needle is locked within the barrel.