Sol-Care® Allergy Syringe Trays, Sol M
Supplier: SOL-MILLENNIUM CANADA, INC. CA
100032IM 100069IM 181023T 181026IDBT 181026T 181027IDBT 181027T 181126IDBT
76582-520CS 550 CAD
76582-520 76582-528 76582-632 76582-634 76582-636 76582-638 76582-640 76582-642
Sol-Care® Allergy Syringe Trays, Sol M
Syringes Syringes, Medical
The Sol-M Allergy Syringe Tray is designed for mixing and administering allergenic extracts.
  • Low Dead Space design reduces waste and allows for more efficient medication delivery
  • Clear, bold graduations for accurate filling and dosing
  • Sterile trays for convenient filling of allergenic extracts
The SOL-M™ Allergy Syringes have transparent barrels with bold, easy to read graduations that allow for accurate filling and dosing. They feature a low dead space design and are packaged in sterile convenience trays.