Sol-M® Blood Collection Tube Holders
Supplier: SOL-MILLENNIUM CANADA, INC. CA
BCTH002 110201050001 110201050003
76582-746BX 80.41 CAD
76582-746 76582-478 76582-480
Sol-M® Blood Collection Tube Holders
Blood Collection Tube Holders
Tube holders for clean blood collection.
Tube holder with Luer lock adapter: Designed for collecting specimens from IV lines or for collecting urine specimens, and is compatible with IV sets, ports, and tubing according to ANSI standards.
Blood transfer holder: Designed to work in a variety of functions by securely connecting to female Luer connections.
Blood collection tube holder: Compatible with standard multi-sample needles and blood collection tubes.