Sol-Care® Safety Multi-Sample Needles, Sol M
Supplier: SOL-MILLENNIUM CANADA, INC. CA
SMSN21125 SMSN22125 SMSNH21125 SMSNH22125
76582-900BX 38.77 CAD
76582-900 76582-902 76582-904 76582-906
Sol-Care® Safety Multi-Sample Needles, Sol M
Syringe Needles
The Sol-Care Safety Multi-Sample Blood Collection Needles are designed with a safety shield that completely covers and locks over the needle after use to protect against needlestick injuries.
  • Safety shield aligns with a bevel up needle orientation, allowing for low-angle penetration
  • Pivoting safety shield with audible click that indicates it is locked in place
  • Safety mechanism is designed for one-handed activation
  • Safety shield is color coded to reduce potential errors
Compatible with all standard holders.