Sol-M® Multi-Sample Needles
Supplier: SOL-MILLENNIUM CANADA, INC. CA
110201020002 110201020003 110201020004 110201020005 110201020006 110201020007
76582-452BX 25.01 CAD
76582-452 76582-438 76582-440 76582-442 76582-444 76582-446 76582-448 76582-450
Sol-M® Multi-Sample Needles
Syringe Needles
Sol-M Multi-Sample Blood Collection Needles allows for the collection of blood into multiple vacuum collection tubes during a single venipuncture.
  • Black dot on multi-sample needle hub ensures bevel up orientation
  • Low-angle bevel design allows easy penetration
  • Color-coded caps and hubs allow easy identification of needle gauge size
  • Premium rubber sheathed needle point for maximum multi-puncture sealing capability
Compatible with all standard holders.
Order Now
SPECIFICATIONS
Environmentally Preferable