Eisco Glass Volumetric Flasks, Class B
Supplier: Wards
CH0450J CH0450G CH0450B CH0450H CH0450D CH0450I CH0450F CH0450E CH0450A CH0450C
470343-722EA 83.25 CAD
470343-722 470343-742 470343-890 470344-526 470344-614 470345-042 470345-240 470345-460 470345-644 470345-738
Eisco Glass Volumetric Flasks, Class B
Flasks Volumetric Flasks
Class B volumetric flasks without stoppers are made from borosilicate 3.3 glass and feature a graduation mark, blue enamel markings, and beaded rims.
  • Made of high quality, heavy duty, borosilicate 3.3 glass for excellent durability and chemical resistance
  • Class B tolerances conform with DIN ISO 1042 standards for accuracy
  • Easy-to-read blue enamel graduations and specifications for contrast against clear liquids
  • Fire-polished beaded rims are chip and crack resistant
Calibrated to contain.
Eisco labs borosilicate glass has the following heatMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable