Eisco Glass Volumetric Flasks with Glass Stopper, ASTM Class A
Supplier: Wards
CH0441CWT CH0441A01 CH0441BWT CH0441D CH0441FWT CH0441HWT CH0441A02 CH0441GWT CH0441AWT CH0441EWT CH0441H CH0441DWT CH0441A02WT
470343-942EA 21.95 CAD
470343-942 470343-958 470344-210 470344-626 470344-696 470344-744 470344-994 470345-114 470345-126 470345-420 470345-604 470345-992 470346-050
Eisco Glass Volumetric Flasks with Glass Stopper, ASTM Class A
Flasks Volumetric Flasks
ASTM Class A volumetric flasks are made from borosilicate 3.3 glass and feature a graduation mark, solid glass pennyhead stopper, and blue or white enamel markings.
  • Made of high quality, heavy duty, borosilicate 3.3 glass for excellent durability and chemical resistance
  • Class A tolerances conform with ASTM E288 standards for accuracy
  • Fitted with a solid glass pennyhead stopper
  • Available with blue or white enamel markings and specifications
Calibrated to contain.
Eisco labs borosilicate glass has the following heat tolerances:
- Strain point 515 °C
- Annealing point 565 °C
- Softening point 820 °C