Eisco Amber Glass Volumetric Flasks with Plastic Stopper, Class A
Supplier: Wards
CH0451I CH0451H CH0451A CH0451B CH0451E CH0451C CH0451J CH0451G CH0451D CH0451F
470343-894EA 74.9 CAD
470343-894 470344-248 470344-294 470344-616 470344-958 470345-376 470345-448 470345-650 470345-940 470346-078
Eisco Amber Glass Volumetric Flasks with Plastic Stopper, Class A
Flasks Volumetric Flasks
Class A Amber volumetric flasks are made from borosilicate 3.3 glass and feature a graduation mark, polypropylene stopper, and white enamel markings.
  • Made of high quality, heavy duty, borosilicate 3.3 glass for excellent durability and chemical resistance
  • Amber color suitable for light-sensitive chemicals
  • Class A tolerances conform with DIN ISO 1042 standards for accuracy
  • Fitted with a chemically-resistant polypropylene flat-top stopper
  • Easy-to-read white enamel markings and specifications
Calibrated to contain.
Eisco Labs borosilicate glass has the following heat tolerances:
- Strain point 515 °C
- Annealing point 565 °C
- Softening point 820 °C