Eisco Glass Volumetric Flasks with Plastic Stopper, Class B
Supplier: Wards
CH0448D CH0448E CH0448A CH0448H CH0448F CH0448B CH0448J CH0448I CH0448G
470343-696EA 13.6 CAD
470343-696 470343-748 470343-884 470344-152 470344-288 470344-418 470345-720 470345-776 470346-100
Eisco Glass Volumetric Flasks with Plastic Stopper, Class B
Flasks Volumetric Flasks
Class B Volumetric flasks are made from borosilicate 3.3 glass and feature a graduation mark, polypropylene stopper, and blue enamel markings.
  • Made of high quality, heavy duty, borosilicate 3.3 glass for excellent durability and chemical resistance
  • Class B tolerances conform with DIN ISO 1042 standards for accuracy
  • Fitted with a chemically-resistant polypropylene flat-top stopper
  • Easy-to-read blue enamel graduations and specifications for contrast against clear liquids
Calibrated to contain.
Eisco Labs borosilicate glass has the following heat tolerances:
- Strain point 515 °C
- Annealing point 565 °C
- Softening point 820 °C