Eisco Glass Volumetric Flasks with Snap Cap, ASTM Class A
Supplier: Wards
CH0443EWT CH0443DWT CH0443F CH0443C CH0443GWT CH0443BWT CH0443CWT CH0443B CH0443D CH0443E CH0443A01WT CH0443A CH0443A01 CH0443FWT CH0443G CH0443AWT
470343-570EA 32.6 CAD
470343-570 470343-596 470343-650 470343-692 470344-124 470344-310 470344-388 470345-172 470345-408 470345-516 470345-546 470345-576 470345-766 470345-872 470345-876 470345-902
Eisco Glass Volumetric Flasks with Snap Cap, ASTM Class A
Flasks Volumetric Flasks
ASTM Class A Volumetric flasks are made from borosilicate 3.3 glass and feature a graduation mark, polypropylene snap cap, and blue or white enamel markings.
  • Made of high quality, heavy duty, borosilicate 3.3 glass for excellent durability and chemical resistance
  • Class A tolerances conform with ASTM E288 standards for accuracy
  • Fitted with a polypropylene snap cap to prevent spillage
  • Available with blue or white enamel markings and specifications
Calibrated to contain.
Eisco Labs borosilicate glass has the following heat tolerances:
- Strain point 515 °C
- Annealing point 565 °C
- Softening point 820 °C