Duo-i Dual Systems, Ultrapure (18.2 MΩ-cm) and DI Lab Water, Avidity Science

Supplier: AVIDITY SCIENCE
7120-4500-473 7120-4500-493 7120-4500-433 7120-4500-443
76499-370EA 27380.93 CAD
76499-370 76499-372 76509-500 76509-502 76533-356
Duo-i Dual Systems, Ultrapure (18.2 MΩ-cm) and DI Lab Water, Avidity Science
Water Purification Systems Pure and Ultra Pure Water Systems
Two-In-One system provides DI or RO water to a storage tank, and capable of dispensing Type 1 and Type 2 or Type 3 water from dispense guns (depending on model variation).

  • Fast, volumetric and variable dispense up to 2 L/min
  • Ability to dispense Type 1 and Type 2 water (optional) from remote dispenser(s)
  • Make-up flow rate of 10 L/h
  • Unique twin pass RO technology ensures enhanced DI cartridge capacity
  • Data capture with printable option
  • Integrated water leakage protection
  • Easy to change consumables with traceability and data tags
  • Interactive 7" touch screen
  • Color coded and adjustable dispensing stations with up to two remote dispensing options
  • Choice of storage reservoir size - 35, 70, or 105 L

The innovative and unique Duo-i provides two water qualities within one easy to use system. Save on space, initial outlay and consumable costs. Type 2 or 3 water (based on model) is available from the reservoir while Type 1 water is ready to dispense from the remote dispenser. Additional dispense guns allow dispensing of water from reservoir. This model filters water into a tank at 10, 20, or 30 L per hour (based on model) which can be dispensed from a 35, 70, or 105 L reservoir.

Application: Duo-i II.I is suitable for Type 2 water usage. Type 2 water is used for buffer and media production, sample and reagent diluent, general chemistry, protein electrophoresis, histology, cytology, and spectrophotometry. Duo-i III.I Type 3 water is suitable to feed an ultrapure water machine, glass washer, humidity/stability chambers and other laboratory equipment. Duo-i II.I and III.I is also suitable for Type 1 water usage. Type 1 water is used for molecular biology, electrophoresis, cell and tissue culture, sequencing, HPLC, genomics, pharmacology, and any other sensitive applications.

Certifications: Manufactured to ISO 9001:2015 standard. Certified to cUL, and IEC 61010-1:2010+A1.

Ordering information: Endotoxin and nuclease-free point of use filter available (see 76337-284). First round of consumables are included with purchase of unit. Ships within 2 to 3 weeks and installed within 4 to 6 weeks from receiving the PO.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR