Eisco Glass Volumetric Flasks with Glass Stopper, ASTM Class B
Supplier: Wards
CH0442CWT CH0442A02 CH0442GWT CH0442A01 CH0442FWT CH0442BWT CH0442D CH0442H CH0442EWT CH0442HWT CH0442A02WT CH0442DWT CH0442AWT
470343-620EA 17.95 CAD
470343-620 470343-664 470343-744 470344-502 470344-672 470344-800 470344-942 470345-654 470345-688 470345-870 470345-934 470345-944 470346-006
Eisco Glass Volumetric Flasks with Glass Stopper, ASTM Class B
Flasks Volumetric Flasks
ASTM Class B Volumetric flasks are made from borosilicate 3.3 glass and feature a graduation mark, solid glass penny head stopper, and blue or white enamel markings.
  • Made of high quality, heavy duty, borosilicate 3.3 glass for excellent durability and chemical resistance
  • Class B tolerances conform with ASTM E288 standards for accuracy
  • Fitted with a solid glass penny head stopper
  • Available with blue or white enamel markings and specifications
Calibrated to contain.
Eisco Labs borosilicate glass has the following heat tolerances:
- Strain Point 515 °C
- Annealing Point 565 °C
- Softening Point 820 °C