Sol M® Shoe Covers, Sol-Millennium
Supplier: SOL-MILLENNIUM CANADA, INC. CA
SC002U SC002XL SC004U SC004XL
76582-842CS 176.83 CAD
76582-842 76582-844 76582-846 76582-848 76582-850 76582-852 76582-854
Sol M® Shoe Covers, Sol-Millennium
Disposable Garments
Shoe covers are durable and available in various weights to accommodate specific protection requirements.
  • Medium weight or heavy weight options
  • Anti-skid traction
  • Elastic cuffs
Shoe covers are made from durable SMS material that comes in both medium weight and heavy weight formats. Anti-skid bands provide traction on the bottom of the covers. Intended to be worn over shoes to prevent the spread of contaminants.