Iron Wire Gauze with Ceramic Center
Supplier: Wards
CH0525B CH0525A
470343-988EA 47.1 CAD
470343-988 470344-476
Iron Wire Gauze with Ceramic Center
Wire
Pack of ten iron wire gauze with ceramic center.
  • Comes in a pack of 10
  • Ceramic centers measure 3" or 4" in diameter
  • High quality and extra strong
  • Free of hazardous materials