Iron Wire Gauze
Supplier: Wards
CH0526B CH0526A
470344-268EA 35.1 CAD
470344-268 470344-488
Iron Wire Gauze
Wire
Iron wire gauze (6×6") is an invaluable component to any chemistry lab.
  • Comes in a pack of 10 gauzes
  • Long lasting, heat and tear resistant iron
  • Free from harmful chemicals and asbestos
Sheet of crosshatched wire used to support glassware during heating. Galvanized iron wire provides a long-life expectancy and is resistant to tearing over time.