CanDo® Exercise Tubing
Supplier: Fabrication Enterprises
10-5724 10-5723 10-5720 10-5721 10-5727 10-5722 10-5726 10-5725
470348-040EA 165.1 CAD
470348-040 470348-012 470348-044 470348-054 470348-028 470348-016 470348-048 470348-004
CanDo® Exercise Tubing
Educational Materials Physical Education Supplies
Bulk rolls for easy length adjustment.
  • Latex-free
  • Lightweight, portable, and durable
CanDo® high-quality, latex-free resistive exercise tubing has been used in clinics for rehabilitation, conditioning, and training since 1987. It has become a staple of many physical therapy and athletic training programs. CanDo® exercise tubing is an economic solution for upper and lower body exercise. Effective when used alone, or with handles and anchors. Exercise tubing is color-coded according to resistance. Simply unroll desired length of exercise tubing and cut with scissors. CanDo® is licensed to use the Thera-Band color sequence so resistance level is easily identified. Tubing is available in 8 resistance levels.