Backyard Brains® Human Spikerbox
Supplier: BACKYARD BRAINS, INC.
BYBP0088 BYBP0089 BYBP0069
470315-130EA 116.35 CAD
470315-130 470315-132 470343-178
Backyard Brains® Human Spikerbox
Educational Materials Biology Educational Materials Physiology Learning Activities
New and improved.
  • Connect via USB or audio cable to free recording software on any device
  • Record from multiple muscles at the same time
  • Add the 'reflex hammer' to precisely measure reflexes like the 'knee-jerk'
  • Add the 'reaction timer' to use muscle contractions to measure reaction times to light and sounds
One box to rule them all! Reveal and record the electrical activity hidden within your muscles, heart, brain, eyes and more! This easy-to-use kit allows you to record EMG, EKG, EEG, and EOG signals, giving you the power to perform human physiologyMore Product Information