LIQUIPORT™ NF100 and NF1.100 Series Diaphragm Liquid Pumps, KNF
Supplier: KNF Neuberger Inc.
179090/153257 179091/153257 179092/153257 179096/153257 179097/153257 179098/153257 179093/153257 179094/153257 179095/153257 179099/153257 179100/153257 179101/153257
89204-480EA 2782.85 CAD
89204-480 89204-482 89204-484 89204-486 89204-488 89204-490 89204-504 89204-506 89204-508 89204-510 89204-512 89204-514
LIQUIPORT™ NF100 and NF1.100 Series Diaphragm Liquid Pumps, KNF
Pumps Diaphragm Pumps
Chemically resistant liquid transfer diaphragm pump. Offers wetted material combinations for optimal chemical resistance. Manual control of flow rate with LED indicator. For convenient transfer and dosing of neutral or corrosive liquids. Option for analog and impulse control (RC).
  • Flow rate of 0.2 to 1.3 L/min
  • Premium corrosion resistance
  • Manual front-panel flow rate control
  • Pressure limit protection
  • No metal wetted parts to corrode
  • Reliable splash-proof housing
  • Models are offered with either PP, PVDF or PTFE pump head material depending on the liquid
More Product Information