LIQUIPORT™ NF300 and NF1.300 Series Diaphragm Liquid Pumps, KNF
Supplier: KNF Neuberger Inc.
179102/153257 179103/153257 179104/153257 179108/153257 179109/153257 179110/153257 179105/153257 179106/153257 179107/153257 179111/153257 179112/153257 179113/153257
89204-492EA 2590.55 CAD
89204-492 89204-494 89204-496 89204-498 89204-500 89204-502 89204-516 89204-518 89204-520 89204-522 89204-524 89204-526
LIQUIPORT™ NF300 and NF1.300 Series Diaphragm Liquid Pumps, KNF
Pumps Diaphragm Pumps
Chemically resistant liquid transfer diaphragm pump. Offers wetted material combinations for optimal chemical resistance. Manual control of flow rate with LED indicator. For convenient transfer and dosing of neutral or corrosive liquids. Option for analog and impulse control (RC).
  • Premium corrosion resistance
  • Manual front-panel flowrate control
  • Pressure limit protection
  • No metal wetted parts to corrode
  • Reliable splash-proof housing
  • Models are offered with either PP, PVDF or PTFE pump head material depending on the liquid being transferred
  • All models
More Product Information