LABOPORT™ UN820 Vacuum Diaphragm Pump, KNF
Supplier: KNF Neuberger Inc.
309577/026366 209642/029940
14224-132EA 4785.98 CAD
14224-132 26678-000 26678-012
LABOPORT™ UN820 Vacuum Diaphragm Pump, KNF
Pumps Vacuum Diaphragm Pumps
Chemically resistant, oil-free diaphragm vacuum pump used for a wide range of laboratory applications. Especially suited for handling aggressive vapors, it features PTFE heads, PTFE-coated diaphragms and FFPM valves.
  • Handle corrosive and aggressive vapors without damage
  • 100% Oil-free transfer
  • Highly compatible with vapours and condensation
  • Chemically resistant
  • Suitable for highly corrosive gases and vapours
  • High level of gas tightness
Pure transferring and evacuation of gases.