Axygen® Sealing Mats for Assay and Storage Microplates, Corning
Supplier: Corning
AM-2ML-RD-S AM-384-DW-SQ AM-2ML-RD AM-2ML-SQ AM-750UL-RD AM-500UL-RD AM-24-SQ
10011-946CS 1542.25 CAD
10011-946 10011-124 10011-126 10011-128 47734-658 47743-826 89179-610
Axygen® Sealing Mats for Assay and Storage Microplates, Corning
Seals Microplate Seals
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications.
  • Minimize evaporation
  • Made of research-grade silicone
  • Autoclavable
They can be reused following a bleach wash and ethanol rinse protocol.