Drying and Heating Chambers with Forced Convection, FP Series, Binder
Supplier: Binder
9010-0367 9010-0368 9010-0369 9010-0370
76627-908EA 6823.39 CAD
76627-908 76627-910 76627-912 76627-914
Drying and Heating Chambers with Forced Convection, FP Series, Binder
Ovens Standard Ovens
The BINDER FP series of material test chambers with mechanical convection and program functions provide reliably short drying times and particularly fast heating – even for chambers under full loads.
  • Uniform drying conditions thanks to APT.line™ technology
  • Identical test conditions throughout the chamber interior independent of sample size and quantity
  • Outstanding thermal insulation saves operation costs
  • Advanced programming options
The ovens have a temperature range of +10 °C above ambient temperature up to +300 °C, they provide high temperatureMore Product Information