Axygen® Automation-Compatible PCR Sealing Mats, Corning
Supplier: Corning
AM-384-PCR-RD AM-96-PCR-RD
10011-122CS 1231.87 CAD
10011-122 10011-130
Axygen® Automation-Compatible PCR Sealing Mats, Corning
Seals Microplate Seals
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications.
  • Minimize evaporation
  • Made of research-grade silicone
  • Autoclavable
They can be reused following a bleach wash and ethanol rinse protocol.