Medical Chemical Corporation Basic Fuchsin, Grams Counterstain, Hardy Diagnostics
Supplier: Hardy Diagnostics
BF008 BF008
CA76591-154EA 24.63 CAD
76591-154 CA76591-154
Medical Chemical Corporation Basic Fuchsin, Grams Counterstain, Hardy Diagnostics
Stains Microbiology Stains
Basic fuchsin a stronger more intense counterstain, that produces a darker red color
Works especially well with anaerobes and other difficult to stain organisms.
Delivery information: Ship ground.