AssureWear™ VersaGown® Isolation Gown with Flexneck™ Technology, Medicom
Supplier: Medicom
A69961 A69962 A69963
CA76545-390CS 187.72 CAD
CA76545-390 CA76545-392 CA76545-394
AssureWear™ VersaGown® Isolation Gown with Flexneck™ Technology, Medicom
Disposable Garments
Patented Flexneck™ technology for fast and easy donning and removal.
  • VersaGown® with Flexneck™ technology significantly reduces cross-contamination
  • Designed using clinician feedback and independent clinical testing
  • Provides nearly two-fold reduction in contamination of hand and wrist areas
  • Helps ensure instant compliance, even without training
  • Innovative design allows freedom of movement without compromising protection
These three layers SMS isolation gowns feature thumb loop cuffs and belt on front. Patented Flexneck™ technology allows fast and easyMore Product Information