Ocean Pacific® NeoNatural™ Polychloroprene Medical Examination Gloves, Powder-Free
Supplier: Medicom
1228-E 1228-A 1228-B 1228-C 1228-D 1228-E
76579-714CS 270.45 CAD
76579-714 CA76579-706 CA76579-708 CA76579-710 CA76579-712 CA76579-714
Ocean Pacific® NeoNatural™ Polychloroprene Medical Examination Gloves, Powder-Free
Gloves
Non sterile, single use, medical examination gloves are intended to prevent transmission of a wide variety of diseases to both patients and health care personnel. Specifically, gloves are intended to act as a barrier between the health care provider and patients to prevent contamination between patient and caregiver.
  • Textured fingertips
  • Chemo-tested and chemical resistant
  • Strength and durability: Synthetic rubber material with a high tensile strength, helps prevent gloves from tearing during donning and extended wear
  • Comfortable fit: Offers softness and flexibility similar to latex gloves, ensuring optimal comfort and conforms to hands helping to prevent fatigue
  • Chemical resistance provides strong chemical resistance for consistent, reliable protection when handling chemical substances
  • Latex-free: Not made with natural rubber latex, so they are safe for professionals and patients with a latex allergy or sensitivity
Combines the softness of latex with the strength of nitrile for reliable protection without sacrificing comfort or dexterity.
Color: Emerald (Green)
Certifications:
Meets ASTM D 6977-04.
Packaging:
100 gloves/box, 10 boxes/case, 1000 gloves/case.