Corning® External Thread Cryogenic Vials, Corning
Supplier: Corning
430658 430659 430661 430662 430663
66021-972CS 663.22 CAD
66021-972 66021-974 66021-976 66021-978 66021-980
Corning® External Thread Cryogenic Vials, Corning
Vials
These vials are manufactured from polypropylene to withstand temperatures down to –196 °C.
  • Black graduations with large marking spot
  • Vials have a silicone washer for a secure seal
  • 2 ml self-standing vials are available in five cap colors.
  • Vials may be color coded with inserts
  • Free foam rack with each case
  • RNase-/DNase-free
  • Sterile
  • Nonpyrogenic
Self-standing vials have a special base design allowing them to be locked into cryogenic rack and tray (Cat. No. 431131) for single-handed manipulation.
Caution: Do not use cryogenic vials for storage in theMore Product Information