Eisco Plastic Conical Flasks with Screw Cap
Supplier: Wards
CH0431C CH0431B CH0431A
470344-122EA 14.85 CAD
470344-122 470345-050 470345-666
Eisco Plastic Conical Flasks with Screw Cap
Flasks Erlenmeyer Flasks
Polypropylene conical flasks with screw caps and no graduations.
  • Made from high-quality polypropylene
  • Highly chemical resistant, microwavable and autoclavable at 121 °C
  • Tight-fitting screw caps for transportation and storage of contents
  • Wide neck for easy filling and cleaning
  • Translucent body makes it easy to see liquid level in the flask
Conical flasks with a wide neck and a screw cap. The natural translucent color makes it easy to see any liquid level in the bottle. Made from easy-to-clean high-quality polypropylene. Highly chemical resistant, microwavable and autoclavable at 121 °C.