VWR® Premium Thickness Nitrile Gloves
Supplier: VWR International
76582-346 76582-340 76582-344 76582-348 76582-338
76582-346BX 32.26 CAD
76582-346 76582-340 76582-344 76582-348 76582-338
VWR® Premium Thickness Nitrile Gloves
Gloves
Ambidextrous 6 mil gloves are extra-soft and comfortable for everyday use. They protect hands from a broad range of chemicals.
  • 6 mil finger thickness
  • Nonsterile
  • Latex-free
  • Extra-soft and comfortable
  • Powder-free
  • Beaded cuff allows easy donning
Ideal for use in industrial and laboratory applications. Made of premium compounded nitrile for high manufacturing quality measures and product consistency. Gloves protect against biohazards and a broad range of chemicals. Composition is 100% latex-free to eliminate adverse reactions associated with naturalMore Product Information