VWR® Extended Cuff Nitrile Gloves
Supplier: VWR International
76582-334 76582-350 76582-336 76582-342
76582-334BX 29.97 CAD
76582-334 76582-350 76582-336 76582-342
VWR® Extended Cuff Nitrile Gloves
Gloves
Ambidextrous gloves with extended cuff are extra-soft and comfortable for everyday use. They protect hands from a broad range of chemicals.
  • 12" extended cuff
  • Nonsterile
  • Latex-free
  • Extra-soft and comfortable
  • Powder-free
  • Beaded cuff allows easy donning
Ideal for use in industrial and laboratory applications. Made of premium compounded nitrile for high manufacturing quality measures and product consistency. Gloves protect against biohazards and a broad range of chemicals. Composition is 100% latex-free to eliminate adverse reactions associated with natural rubber latex proteins. Full textured gloves feature rolled, beaded cuffs.
Certifications: Gloves meet and exceed ASTM D6319 specification for Nitrile Examination Gloves for Medical Application.
Packaging: Gloves are packaged 100 per box, 10 boxes per case.