VWR® dNTPs

Supplier: VWR International

76620-542 76620-544 76620-546 76620-548
76620-542EA 457.55 CAD
76620-542 76620-544 76620-546 76620-548
VWR® dNTPs
Nucleic Acid Reagents Nucleotides

Ready to use molecular biology grade dNTP mixes and dNTP sets.


  • Available as pre-mixed 40 or 100 mM solutions , or as sets of individual 100 mM dNTP solutions
  • Both pre-mixed and sets have been functionally tested in PCR
  • Purity >99% by HPLC
  • Supplied in solution at pH 7.3 to 7.5


The dNTP mix is designed to save hands-on time for researchers, and reduce the possibility of contamination by reducing pipetting. The dNTP mix and dNTP solutions are convenient for use in DNA polymerization reactions, DNA labelling and sequencing processes.


Caution: For Research Use Only. Not for use in diagnostic procedures.

Order Now

SPECIFICATIONS

Environmentally Preferable
Application

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR