VWR® GC Tempase Hot Start DNA Polymerase Master Mixes
Supplier: VWR International
76620-500 76620-524 76620-526 76620-528
76620-500EA 198.08 CAD
76620-500 76620-524 76620-526 76620-528
VWR® GC Tempase Hot Start DNA Polymerase Master Mixes
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Ready to use GC TEMPase 2X Master Mixes I and II are designed for amplification of GC-rich sequences.
  • High success rate with amplification of GC-rich DNA
  • High sensitivity, specificity and product yield
  • Diminished formation of non-specific products
  • Reaction set-up at room temperature
  • Processes up to 4 kb
The GC TEMPase 2X master mixes consist of TEMPase Hot Start DNA Polymerase combined with an optimized GC buffer system (GC buffer I or II), dNTPs and MgCl2. Just add DNA template and primers to carry out PCR.
GC-Rich DNA Target Kit consists of TEMPase Hot Start DNA polymerase and seperate tubes with GC buffer I and II. The kit is designed for initial testing before using one of the GC-TEMPase 2X Master Mixes.
Long term storage at -20 °C. Product expiry at -20 °C is stated on the label. Optional: Store at +4 °C for up to 6 months.
Caution: For Research Use Only. Not for use in diagnostic procedures.