VWR® Fast HiFi DNA Polymerase

Supplier: VWR International

76620-574 76620-576 76620-578 76620-580
76620-574EA 210.12 CAD
76620-574 76620-576 76620-578 76620-580
VWR® Fast HiFi DNA Polymerase
Nucleic Acid Reagents End-point PCR Enzymes and Kits

Fast HiFi DNA Polymerase (2 U/µl) is a proofreading DNA polymerase with the following features: High fidelity >60X Taq DNA Polymerase, ability to amplify problematic DNA targets, such as those with low to high GC content, and ability to perform amplification on long DNA targets.


  • High fidelity: >60X Taq Fidelity
  • 3’→5’ proofreading exonuclease activity
  • Processes up to 18 kb (human gDNA) and 25 kb (λDNA)
  • Good coverage on DNA templates with low to high GC content
  • Fast extension rate: 10 s/kb


Fast HiFi DNA Polymerase: 2 Units/μl

Fast HiFi DNA Polymerase exhibits both 5'→3' DNA polymerase activity and 3'→5' proofreading exonuclease activity enabling this polymerase to correct base pair mismatches. It is supported with 5X Fast HiFi Buffer allowing robust amplification on DNA targets with low to high GC content and long DNA targets.

For difficult amplicons, such as GC-rich DNA samples, those with complex secondary structures or long amplicons, the addition of 1 to 2 M Betaine Enhancer Solution is recommended.

Fast HiFi DNA Polymerase is recommended for applications which require extremely high fidelity, such as cloning/subcloning, NGS applications, and mutagenesis.

Sample types: gDNA, plasmid DNA, bacterial DNA, λDNA, and other DNA templates.

Long term storage at -20 °C. Product expiry at -20 °C is stated on the label. Optional: Store at +4 °C for up to 6 months.

For more convenient handling, Fast HiFi DNA Polymerase is also available as a 2X master mix.


Caution: For Research Use Only. Not for use in diagnostic procedures.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR