VWR® TAQ Plus DNA Polymerase Master Mix
Supplier: VWR International
76620-536 76620-538 76620-540 76620-534
76620-536EA 339.32 CAD
76620-536 76620-538 76620-540 76620-534
VWR® TAQ Plus DNA Polymerase Master Mix
Nucleic Acid Reagents End-point PCR Enzymes and Kits
VWR® Taq Plus is an optimized format of Taq DNA polymerase master mix and, therefore, is a good alternative to Taq DNA polymerase and Taq DNA polymerase master mix.
  • Ready to use 2X master mix for greater convenience
  • Significantly reduced set-up time
  • Increased specificity compared to Taq DNA polymerase master mix
  • Reduced risk of contamination
  • Processes up to 4 kb
Ensures increased specificity and improved PCR performance, even on difficult templates. These reagents are suitable for standard PCR applications, screening and high-throughput testing.
This PCR master mix is composed of Taq DNA polymerase and an optimized buffer system, including dNTPs and magnesium chloride (1.5 mM).
Long term storage at -20 °C. Product expiry at -20 °C is stated on the label. Optional: Store at +4 °C for up to 6 months.
Caution: For Research Use Only. Not for use in diagnostic procedures.