VWR® RED Taq® DNA Polymerase Master Mix
Supplier: VWR International
VWR® Red Taq DNA Polymerase Master Mix, which also contains an inert red dye, can be directly loaded onto an agarose gel without addition of electrophoresis loading buffers.
- All-in-one master mix for great convenience
- Timesaving reaction setup
- Direct loading onto agarose gel
- Easy visualisation of pipetting
- Red tracking dye
- Increased reproducibility
- High product yield
- Processes up to 5 kb
Red Taq DNA Polymerase master mixes are a good alternative to VWR® Taq DNA Polymerase. The inert red dye and stabilizer are extra features, which allow direct loading onto your agarose gel for analysis.
The Red Taq DNA Polymerase master mixes offer reduced set-up time and eliminate the risk of contamination of reaction components. Fewer handling steps lead to better reproducibility, which makes master mixes suitable for standard PCR applications, screening and high-throughput testing.
Red Taq DNA Polymerase Master Mix is a ready-to-use 1.1X or 2X reaction mix. Simply add primers, template, and water to carry out primer extensions and other molecular biology applications.
Long term storage at -20 °C. Product expiry at -20 °C is stated on the label. Optional: Store at +4 °C for up to 6 months.
VWR® Red Taq DNA Polymerase 1.1X and 2X master mixes are available in two final MgCl2 concentrations: 1.5 mM and 2 mM.
Caution: For Research Use Only. Not for use in diagnostic procedures.