Axygen® Deep Well and Assay Plates, Corning
Supplier: Corning
Axygen®
P-96-450V-C-S P-96-450R-C-S P-96-450R-C
490005-358CS 520.8 CAD
490005-358 490005-360 490005-364
Axygen® Deep Well and Assay Plates, Corning
Microplates Deep Well Microplates
Axygen® deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage.
  • 0.6 ml deep well plate
  • ANSI/SLAS 1-2004: Microplates - footprint dimensions
  • Ultra-flat surface ensures proper sealing with heat sealing film
Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to −80 °C.