FlexTech™ Y9258 Cut-Resistant ESD Gloves, Wells Lamont
Supplier: Wells Lamont
Y9258L Y9258M Y9258S Y9258XL Y9258XXL
76557-512DZ 203.08 CAD
76557-512 76557-514 76557-516 76557-520 76557-518
FlexTech™ Y9258 Cut-Resistant ESD Gloves, Wells Lamont
Gloves
The Y9258 gloves offer anti-static properties that rapidly neutralize the electric discharge for the worker and ESD properties that protect product equipment and workpieces. The ultra-lightweight liner provides excellent dexterity with ANSI Cut Level A4 protection to reduce hand injuries, and increased abrasion resistance and durability for prolonged use.
  • ANSI A4 cut resistance
  • ESD and anti-static properties
  • 18 gauge thin liner
  • Foam nitrile palm for grip
  • ANSI Level 4 puncture resistance
  • ANSI Level 4 abrasion resistance
  • ANSI Level 1 heat resistance
  • Touchscreen capable
Specifically designed for use where anti-static or ESD protection is needed.
Certifications: Gloves meet ANSI Level A4 for cut resistance, ANSI Level 4 for puncture resistance, ANSI Level 4 for abrasion resistance, ANS Level 1 for heat resistance.