Eisco Plastic Graduated Cylinders
Supplier: Wards
CH0354C CH0354B CH0354E CH0354F CH0354H CH0354G CH0354A CH0354D
470343-822EA 4.45 CAD
470343-822 470344-148 470344-156 470344-338 470345-324 470345-744 470345-798 470346-074
Eisco Plastic Graduated Cylinders
Cylinders Measuring Cylinders
Economical polypropylene graduated cylinders with an integrated hexagonal base.
  • Made of premium polypropylene plastic with high heat tolerance and chemical resistance
  • Translucent rigid construction and superior optical clarity
  • Easy-to-read, raised graduation lines
  • Spout for easy pouring
  • Hexagonal base provides maximum stability and prevents rolling when placed on side
  • Microwavable and autoclavable at 121 °C
Microwavable and resistant to fatigue for long term durability. High heat tolerance with a temperature range of –20 to +135 °C. Good to excellent chemical resistance.
Packaging: One unit per pack.