Eisco ASTM Glass Graduated Cylinders, Class B
Supplier: Wards
CH0341K CH0341LBL CH0341NBL CH0341KBL CH0341IBL CH0341JBL CH0341MBL CH0341OBL
470344-128EA 15.5 CAD
470344-128 470344-240 470344-530 470345-514 470345-572 470345-662 470346-012 470346-106
Eisco ASTM Glass Graduated Cylinders, Class B
Cylinders Measuring Cylinders
High precision, durability, and chemical resistance.
  • Made of high quality, heavy duty, borosilicate 3.3 glass
  • Conforms with ASTM E1272 Class B tolerance standards
  • Spout for easy pouring
  • Hexagonal base provides maximum stability and prevents rolling when placed on side
  • Easy-to-read graduations and specifications
  • Fitted with a removable, plastic protective collar
Autoclavable, with superior chemical durability. Low temperature gradient - can withstand high temperatures and thermal shocks. Calibrated to contain.
Eisco labs borosilicate glass has theMore Product Information