Ammonia solution 28.0-30.0% NH₃ ACS

Supplier: Ricca Chemical
Danger

RABA0020-2.5D1 RABA0020-500D1
RCRABA00202.5DEA 175.38 CAD
RCRABA00202.5D RCRABA0020500D
Ammonia solution 28.0-30.0% NH₃ ACS
Ammonia solution
Formula: NH₃
Boiling Pt: ~ 36 °C
Melting Pt: ~ 0 °C
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless liquid
Assay 28.0 - 30.0 % NH₃
Carbon Dioxide (CO₂) max 0.002 %
Heavy Metals (as Pb) max 0.5 ppm
Nitrate (NO₃) max 2 ppm
Phosphate (PO₄) max 2 ppm
Sulfate (SO₄) max 2 ppm
Chloride (Cl) max 0.5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR