Potassium hydroxide ≥85% ACS

Supplier: Ricca Chemical
Danger

Synonyms: Potash caustic, Caustic potash

RDCP0500-1000B1 RDCP0500-100B1 RDCP0500-500B1
RCRDCP05001000EA 251.22 CAD
RCRDCP05001000 RCRDCP0500100B RCRDCP0500500B
Potassium hydroxide ≥85% ACS
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: ~1320 °C
Melting Pt: 360
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1813
ADR: 8 II
Merck Index: 12,07806

Order Now

Specification Test Results

Assay min 85 %
Heavy Metals (as Ag) max 0.001 %
Nitrogen Compounds (as N) max 0.001 %
Phosphate (PO₄) max 5 ppm
Potassium Carbonate (K₂CO₃) max 2.0 %
Sulfate (SO₄) max 0.003 %
Sodium (Na) max 0.05 %
Magnesium (Mg) max 0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR